siRNA Details
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107929.2 | NP_001101399.2 | 3 | 574 | ||
XM_006238154.2 | XP_006238216.1 | 4 | 499 | |||
XM_006238155.2 | XP_006238217.1 | 4 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
