siRNA Details
Gene Symbol: Trim14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238060.2 | XP_006238122.1 | 2, 3 | 376 | ||
XM_006238061.2 | XP_006238123.1 | 2, 3 | 281 | |||
XM_006238062.2 | XP_006238124.1 | 2, 3 | 389 | |||
GenBank mRNA | BC166777.1 | - | 2, 3 | 316 | 7934596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
