siRNA Details
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107938.1 | NP_001101408.1 | 7 | 709 | ||
XM_008763673.1 | XP_008761895.1 | 7 | 1041 | |||
GenBank mRNA | BC158699.1 | - | 7 | 697 | 9028317 |
