siRNA Details
Gene Symbol: Klhl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107944.1 | NP_001101414.1 | 1 | 936 | ||
GenBank mRNA | BC168717.1 | - | 1 | 919 | 9102826 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172871, AK033231, BC094556, AK129338, AK049803.
Gene Ontology Categories:
