siRNA Details
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015015.1 | NP_001015015.1 | 3 | 724 | ||
GenBank mRNA | BC092574.1 | - | 3 | 724 | 7113947 |

More Information
Other Species Targeted

Gene Ontology Categories:
