siRNA Details
Gene Symbol: Sgip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763917.1 | XP_008762139.1 | 14 | 1333 | ||
XM_008763918.1 | XP_008762140.1 | 13 | 1248 | |||
XM_008763919.1 | XP_008762141.1 | 16 | 1360 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
