siRNA Details
Gene Symbol: C8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191759.1 | NP_001178688.1 | 7 | 935 | ||
XM_006238495.2 | XP_006238557.1 | 8 | 1255 | |||
GenBank mRNA | BC090023.1 | - | 4 | 460 | ||
FQ211372.1 | - | 1 | 23 |
