siRNA Details
Gene Symbol: Fam151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005558.1 | NP_001005558.1 | 8 | 1285 | ||
GenBank mRNA | BC081977.1 | - | 8 | 1285 | 7127557 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005558.1 | NP_001005558.1 | 8 | 1285 | ||
GenBank mRNA | BC081977.1 | - | 8 | 1285 | 7127557 |