siRNA Details
Gene Symbol: Tmem255a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182822.3 | NP_877974.2 | 8 | 796 | ||
XM_006257479.2 | XP_006257541.1 | 7 | 727 | |||
GenBank mRNA | AY327410.1 | - | 8 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
