siRNA Details
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177931.2 | NP_808792.1 | 15 | 2343 | ||
XM_006238515.2 | XP_006238577.1 | 16 | 2545 | |||
XM_006238516.2 | XP_006238578.1 | 17 | 2658 | |||
GenBank mRNA | AB105378.1 | - | 15 | 2343 |
