siRNA Details
Gene Symbol: Ccdc107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749929.3 | XP_003749977.1 | 5 | 536 | ||
XM_006238135.2 | XP_006238197.1 | 5 | 548 | |||
XM_006238136.2 | XP_006238198.1 | 5 | 544 | |||
XM_006238137.2 | XP_006238199.1 | 5 | 540 | |||
GenBank mRNA | FQ229548.1 | - | 6 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
