siRNA Details
Gene Symbol: Tmem53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107964.1 | NP_001101434.1 | 2, 3 | 275 | ||
XM_006238665.2 | XP_006238727.1 | 2, 3 | 865 | |||
GenBank mRNA | FQ219156.1 | - | 2, 3 | 229 |

More Information
Other Species Targeted

