siRNA Details
Gene Symbol: RGD1564804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107969.1 | NP_001101439.1 | 4 | 373 | ||
GenBank mRNA | FQ209773.1 | - | 4 | 399 |

More Information
Other Species Targeted

