siRNA Details
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107974.1 | NP_001101444.1 | 4 | 758 | ||
GenBank mRNA | FQ211274.1 | - | 499 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107974.1 | NP_001101444.1 | 4 | 758 | ||
GenBank mRNA | FQ211274.1 | - | 499 |