siRNA Details
Gene Symbol: Thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009693.1 | NP_001009693.1 | 5 | 1502 | ||
XM_006238866.2 | XP_006238928.1 | 5 | 1482 | |||
XM_008764021.1 | XP_008762243.1 | 6 | 1601 | |||
XM_008764022.1 | XP_008762244.1 | 5 | 1851 | |||
GenBank mRNA | BC088415.1 | - | 5 | 1502 | 7110766 |
