siRNA Details
Gene Symbol: Pdik1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107984.1 | NP_001101454.1 | 2 | 246 | ||
XM_006239121.2 | XP_006239183.1 | 2 | 784 |

More Information
Other Species Targeted

Gene Ontology Categories:
