siRNA Details
Gene Symbol: Pdik1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107984.1 | NP_001101454.1 | 3 | 996 | ||
XM_006239121.2 | XP_006239183.1 | 3 | 1534 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_146156, NM_001163794, AK164975, AK141566, AK157013, and others.
Gene Ontology Categories:
