siRNA Details
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034081.1 | NP_001029253.1 | 1 | 177 | ||
XM_006239130.2 | XP_006239192.1 | 2 | 275 | |||
XM_008764145.1 | XP_008762367.1 | 3 | 498 | |||
XM_008764146.1 | XP_008762368.1 | 3 | 286 | |||
GenBank mRNA | DQ027002.1 | - | 1 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
