siRNA Details
Gene Symbol: Rap1gap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100713.1 | NP_001094183.1 | 7, 8 | 512 | ||
XM_006239189.1 | XP_006239251.1 | 6, 7 | 589 | |||
XM_006239193.1 | XP_006239255.1 | 6, 7 | 589 | |||
GenBank mRNA | CB581996.1 | - | 7, 8 | 524 |
