siRNA Details
Gene Symbol: Ubr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039026.1 | NP_001034115.1 | 25 | 3477 | ||
XM_006239200.2 | XP_006239262.1 | 25 | 3528 | |||
XM_006239201.2 | XP_006239263.1 | 25 | 3528 | |||
XM_006239202.2 | XP_006239264.1 | 25 | 3528 | |||
XM_006239203.2 | XP_006239265.1 | 25 | 3528 | |||
XM_006239204.2 | XP_006239266.1 | 25 | 3528 | |||
XM_006239205.2 | XP_006239267.1 | 25 | 3528 | |||
XM_006239206.2 | XP_006239268.1 | 25 | 3528 | |||
XM_006239207.2 | XP_006239269.1 | 25 | 3528 | |||
XM_006239208.2 | XP_006239270.1 | 25 | 3528 | |||
GenBank mRNA | AY850927.1 | - | 25 | 3477 |
