siRNA Details
Gene Symbol: Spsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107994.2 | NP_001101464.1 | 2 | 1001 | ||
XM_006239462.2 | XP_006239524.1 | 2 | 1034 | |||
XM_008764257.1 | XP_008762479.1 | 2 | 1247 | |||
XM_008764258.1 | XP_008762480.1 | 1 | 832 | |||
XM_008764259.1 | XP_008762481.1 | 2 | 2978 | |||
GenBank mRNA | BC158603.1 | - | 2 | 1001 | 7133644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
