siRNA Details
Gene Symbol: Cptp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007703.1 | NP_001007704.1 | 3 | 453 | ||
XM_006239576.2 | XP_006239638.1 | 3 | 614 | |||
GenBank mRNA | BC083599.1 | - | 3 | 453 | 7133008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
