siRNA Details
Gene Symbol: Dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108004.1 | NP_001101474.1 | 2 | 260 | ||
XM_006238070.2 | XP_006238132.1 | 2 | 293 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: BC027782, AF088983.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
