siRNA Details
Gene Symbol: Vit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764427.1 | XP_008762649.1 | 4 | 379 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764427.1 | XP_008762649.1 | 4 | 379 |