siRNA Details
Gene Symbol: Supt7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108010.2 | NP_001101480.1 | 3, 4 | 1001 | ||
GenBank mRNA | BC168944.1 | - | 3, 4 | 1001 | 8371512 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028150, AK144989, BC031447, AK012134, AK047063.
Gene Ontology Categories:
