siRNA Details
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750126.3 | XP_003750174.1 | 37 | 5573 | ||
XM_006239862.2 | XP_006239924.1 | 38 | 5672 | |||
XM_006239863.2 | XP_006239925.1 | 37 | 5591 | |||
XM_008764569.1 | XP_008762791.1 | 39 | 5209 | |||
XM_233945.8 | XP_233945.6 | 36 | 5491 |
