siRNA Details
Gene Symbol: Klf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037354.1 | NP_001032431.1 | 3 | 823 | ||
XM_006239984.2 | XP_006240046.1 | 3 | 949 | |||
GenBank mRNA | DQ232764.2 | - | 3 | 823 |

More Information
Other Species Targeted

Gene Ontology Categories:
