siRNA Details
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200045.1 | NP_001186974.1 | 24, 25 | 3274 | ||
XM_008764616.1 | XP_008762838.1 | 23, 24 | 3320 | |||
GenBank mRNA | AB558551.1 | - | 24, 25 | 3274 | ||
FQ225774.1 | - | 19, 20 | 2294 |
