siRNA Details
Gene Symbol: Pnpla8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750150.3 | XP_003750198.1 | 10 | 2493 | ||
GenBank mRNA | FQ211866.1 | - | 5 | 702 | ||
FQ224093.1 | - | 4 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
