siRNA Details
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750171.2 | XP_003750219.1 | 11, 12 | 1901 | ||
XM_006240140.2 | XP_006240202.1 | 11, 12 | 2409 | |||
XM_006240141.2 | XP_006240203.1 | 12, 13 | 2306 | |||
XM_006240142.2 | XP_006240204.1 | 11, 12 | 1971 | |||
XM_006240143.2 | XP_006240205.1 | 11, 12 | 1619 | |||
XM_006240144.2 | XP_006240206.1 | 10, 11 | 1555 | |||
GenBank mRNA | BC166472.1 | - | 11, 12 | 2652 |
