siRNA Details
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199269.1 | NP_954890.1 | 9 | 1960 | ||
XM_008764688.1 | XP_008762910.1 | 8 | 2749 | |||
GenBank mRNA | AY371924.1 | - | 9 | 1960 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199269.1 | NP_954890.1 | 9 | 1960 | ||
XM_008764688.1 | XP_008762910.1 | 8 | 2749 | |||
GenBank mRNA | AY371924.1 | - | 9 | 1960 |