siRNA Details
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199269.1 | NP_954890.1 | 9 | 1922 | ||
XM_008764688.1 | XP_008762910.1 | 8 | 2711 | |||
GenBank mRNA | AY371924.1 | - | 9 | 1922 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001193266, NM_207010, BC137902, BC137901, AY371925, and others.
Gene Ontology Categories:
