siRNA Details
Gene Symbol: Gpr135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181771.1 | NP_861436.1 | 1 | 1073 | ||
GenBank mRNA | AY288430.1 | - | 1 | 1073 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181771.1 | NP_861436.1 | 1 | 1073 | ||
GenBank mRNA | AY288430.1 | - | 1 | 1073 |