siRNA Details
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034929.1 | NP_001030101.1 | 7 | 1161 | ||
XM_006240283.2 | XP_006240345.1 | 7 | 1465 | |||
XM_008764784.1 | XP_008763006.1 | 6 | 1594 | |||
GenBank mRNA | BC105849.1 | - | 7 | 1161 | 7457541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
