siRNA Details
Gene Symbol: Rnf113a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004445.2 | NP_001004445.1 | 1 | 850 | ||
GenBank mRNA | AY363109.1 | - | 1 | 931 | ||
BC100134.1 | - | 1 | 850 | 7133304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
