siRNA Details
Gene Symbol: Gpatch2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134559.1 | NP_001128031.1 | 9 | 1339 | ||
XM_008764794.1 | XP_008763016.1 | 7 | 1253 |

More Information
Other Species Targeted

