siRNA Details
Gene Symbol: Tmem63c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108045.1 | NP_001101515.1 | 22 | 2468 | ||
XM_006240382.2 | XP_006240444.1 | 21 | 2456 | |||
XM_006240383.2 | XP_006240445.1 | 23 | 2610 | |||
XM_006240384.2 | XP_006240446.1 | 22 | 2241 | |||
XM_006240385.2 | XP_006240447.1 | 22 | 2245 | |||
XM_006240386.2 | XP_006240448.1 | 22 | 2291 | |||
XM_006240387.2 | XP_006240449.1 | 22 | 2662 | |||
XM_006240389.2 | XP_006240451.1 | 20 | 2308 | |||
XM_006240390.1 | XP_006240452.1 | 17 | 1646 | |||
XM_008764798.1 | XP_008763020.1 | 22 | 2519 | |||
GenBank mRNA | FQ212398.1 | - | 21 | 2159 |
