siRNA Details
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108046.2 | NP_001101516.1 | 8 | 720 | ||
GenBank mRNA | BC166825.1 | - | 8 | 720 | 8370850 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108046.2 | NP_001101516.1 | 8 | 720 | ||
GenBank mRNA | BC166825.1 | - | 8 | 720 | 8370850 |