siRNA Details
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177933.2 | NP_808794.2 | 18 | 1901 | ||
GenBank mRNA | AF304853.1 | - | 19 | 1915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
