siRNA Details
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005888.1 | NP_001005888.1 | 14 | 1548 | ||
XM_008764799.1 | XP_008763021.1 | 14 | 1984 | |||
GenBank mRNA | AY386370.1 | - | 14 | 1548 | ||
FQ223346.1 | - | 14 | 1567 | |||
FQ228993.1 | - | 14 | 1567 |
