siRNA Details
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108050.2 | NP_001101520.2 | 13 | 2541 | ||
NM_001271174.1 | NP_001258103.1 | 13 | 2838 | |||
GenBank mRNA | DY321166.1 | - | 4 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
