siRNA Details
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108050.2 | NP_001101520.2 | 11 | 2200 | ||
NM_001271174.1 | NP_001258103.1 | 11 | 2497 | |||
GenBank mRNA | DY321166.1 | - | 2 | 237 | ||
EV776435.1 | - | 6 | 686 |

More Information
Other Species Targeted

Gene Ontology Categories:
