siRNA Details
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108050.2 | NP_001101520.2 | 7 | 1629 | ||
NM_001271174.1 | NP_001258103.1 | 7 | 1887 | |||
GenBank mRNA | EV776435.1 | - | 2 | 76 | ||
FQ087395.1 | - | 4 | 607 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001160214, NM_211355, AK173331, AK152480, AK051239, and others.
Gene Ontology Categories:
