siRNA Details
Gene Symbol: Trip11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191772.1 | NP_001178701.1 | 19, 20 | 5459 | ||
XM_006240469.2 | XP_006240531.1 | 18, 19 | 6059 | |||
XM_006240470.2 | XP_006240532.1 | 18, 19 | 6056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
