siRNA Details
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240471.2 | XP_006240533.1 | 12 | 4434 | ||
XM_006240472.2 | XP_006240534.1 | 12 | 4377 | |||
GenBank mRNA | BC168870.1 | - | 11 | 1247 | 7104573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
