siRNA Details
Gene Symbol: Degs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017457.2 | NP_001017457.1 | 2 | 574 | ||
GenBank mRNA | AJ938082.1 | - | 2 | 507 | ||
BC098701.1 | - | 2 | 574 | 7387212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
