siRNA Details
Gene Symbol: LOC102554884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240612.2 | XP_006240674.1 | 10 | 1049 | ||
XM_008764974.1 | XP_008763196.1 | 8 | 1938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
