siRNA Details
Gene Symbol: Rd3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134560.1 | NP_001128032.1 | 3 | 904 | ||
GenBank mRNA | BC158708.1 | - | 3 | 889 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
