siRNA Details
Gene Symbol: Tmem121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001079243.5 | XP_001079243.2 | 2 | 803 | ||
XM_006240674.2 | XP_006240736.1 | 2 | 779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
