siRNA Details
Gene Symbol: Gulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013171.2 | NP_001013189.1 | 6, 7 | 503 | ||
XM_006244804.2 | XP_006244866.1 | 5, 6 | 445 | |||
XM_008767023.1 | XP_008765245.1 | 5, 6 | 447 | |||
XM_008767024.1 | XP_008765246.1 | 5, 6 | 445 | |||
GenBank mRNA | BC087053.1 | - | 6, 7 | 503 | 7123670 | |
DQ668344.1 | - | 6, 7 | 487 |

More Information
Other Species Targeted

Gene Ontology Categories:
